ENA390347249 Enamine

(2R)-2-(benzylamino)propanoic acid

  • MDL number MFCD08443549
  • Cas Number : 65491-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125858
Availability details for
Quantity
  • Availability
  • Add to Cart