ENA390347252 Enamine

(2S,3S)-3-methyl-2-[(1-methyl-2-oxo-1,2-dihydropyridin-4-yl)formamido]pentanoic acid

  • MDL number MFCD16376871
  • Cas Number : 1807896-11-2
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S,3S)-3-methyl-2-{[(1-methyl-2-oxo-1,2-dihydro-4-pyridinyl)carbonyl]amino}pentanoic acid
Inchi Code 1S/C13H18N2O4/c1-4-8(2)11(13(18)19)14-12(17)9-5-6-15(3)10(16)7-9/h5-8,11H,4H2,1-3H3,(H,14,17)(H,18,19)/t8-,11-/m0/s1
Inchi Key RDYDCVRECMUTNY-KWQFWETISA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-125864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125864
Availability details for
Quantity
  • Availability
  • Add to Cart