ENA390347253 Enamine

1-(1-methyl-2-oxo-1,2-dihydropyridine-4-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD11549921
  • Cas Number : 1309019-70-2
  • Molecular Weight : 250.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125865
Availability details for
Quantity
  • Availability
  • Add to Cart