ENA390347255 Enamine

1-chloro-1,1-difluoro-4-phenylbutan-2-one

  • MDL number MFCD23701803
  • Cas Number : 145299-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124817

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124817
Availability details for
Quantity
  • Availability
  • Add to Cart