ENA390347265 Enamine

2-[4-(2,4-difluorophenoxy)phenyl]ethan-1-amine

  • MDL number MFCD19528540
  • Cas Number : 1432680-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125933
Availability details for
Quantity
  • Availability
  • Add to Cart