ENA390347268 Enamine

3-(tert-butylsulfanyl)propan-1-ol

  • MDL number MFCD12178179
  • Cas Number : 5755-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125939
Availability details for
Quantity
  • Availability
  • Add to Cart