ENA390347278 Enamine

ethyl 4-cyclopropyl-5-methyl-4H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD18338217
  • Cas Number : 1342499-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125952
Availability details for
Quantity
  • Availability
  • Add to Cart