ENA390347288 Enamine

4-(4-bromophenyl)benzene-1-sulfonamide

  • MDL number MFCD20230139
  • Cas Number : 454694-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125967
Availability details for
Quantity
  • Availability
  • Add to Cart