ENA390347289 Enamine

2-hydroxy-6-(propan-2-yl)pyridine-4-carboxylic acid

  • MDL number MFCD20642202
  • Cas Number : 90610-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 309...311
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125968

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125968
Availability details for
Quantity
  • Availability
  • Add to Cart