ENA390347290 Enamine

2-[(oxetan-3-yl)amino]ethan-1-ol

  • MDL number MFCD18849303
  • Cas Number : 1343942-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125977
Availability details for
Quantity
  • Availability
  • Add to Cart