ENA390347297 Enamine

6-methyl-1H,2H,3H-pyrido[2,3-b][1,4]oxazine

  • MDL number MFCD20688280
  • Cas Number : 1198154-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126013
Availability details for
Quantity
  • Availability
  • Add to Cart