ENA390347299 Enamine

3,4-diethylbenzoic acid

  • MDL number MFCD20644292
  • Cas Number : 186253-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126015
Availability details for
Quantity
  • Availability
  • Add to Cart