ENA390347304 Enamine

3-{[(4-methoxypyrimidin-2-yl)amino]methyl}benzoic acid

  • MDL number MFCD13166808
  • Cas Number : 1183598-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126022
Availability details for
Quantity
  • Availability
  • Add to Cart