ENA390347305 Enamine

1-{bicyclo[2.2.1]heptan-2-yl}-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD23701916
  • Cas Number : 1432680-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126024

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126024
Availability details for
Quantity
  • Availability
  • Add to Cart