ENA390347308 Enamine

1-(2,4-difluorophenyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202311
  • Cas Number : 1038711-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126028
Availability details for
Quantity
  • Availability
  • Add to Cart