ENA390347312 Enamine

3,4-diethylaniline hydrochloride

  • MDL number MFCD18827535
  • Cas Number : 1071678-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126034
Availability details for
Quantity
  • Availability
  • Add to Cart