ENA390347313 Enamine

5-phenyl-1H-imidazol-2-amine

  • MDL number MFCD03308784
  • Cas Number : 6775-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126036

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126036
Availability details for
Quantity
  • Availability
  • Add to Cart