ENA390347314 Enamine

1-(3,3-dimethylpiperazin-1-yl)ethan-1-one

  • MDL number MFCD19285182
  • Cas Number : 1266867-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126037
Availability details for
Quantity
  • Availability
  • Add to Cart