ENA390347316 Enamine

N-benzyl-7-chloroquinolin-4-amine

  • MDL number MFCD07325309
  • Cas Number : 865270-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126041

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126041
Availability details for
Quantity
  • Availability
  • Add to Cart