ENA390347330 Enamine

2-bromo-5-chloropyridin-3-ol

  • MDL number MFCD09835245
  • Cas Number : 127561-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126094

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-126094
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart