ENA390347334 Enamine

1-(3-bromopropanesulfonyl)-4-methylbenzene

  • MDL number MFCD11164524
  • Cas Number : 5755-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126099
Availability details for
Quantity
  • Availability
  • Add to Cart