ENA390347337 Enamine

2-(3-methyl-2-oxo-1,2-dihydropyridin-1-yl)acetic acid

  • MDL number MFCD11901686
  • Cas Number : 1176754-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126104
Availability details for
Quantity
  • Availability
  • Add to Cart