ENA390347338 Enamine

(1-methyl-3-phenyl-1H-pyrazol-4-yl)methanol

  • MDL number MFCD04972644
  • Cas Number : 499770-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126105

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126105
Availability details for
Quantity
  • Availability
  • Add to Cart