ENA390347341 Enamine

ethyl 2-cyano-2-(prop-2-en-1-yl)pent-4-enoate

  • MDL number MFCD23701927

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126109
Availability details for
Quantity
  • Availability
  • Add to Cart