ENA390347355 Enamine

1-carbamimidamido-N-(2,6-difluorophenyl)methanimidamide hydrochloride

  • MDL number MFCD00276372
  • Cas Number : 242475-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126125
Availability details for
Quantity
  • Availability
  • Add to Cart