ENA390347360 Enamine

[3-methoxy-4-(3-phenylpropoxy)phenyl]methanol

  • MDL number MFCD13566860
  • Cas Number : 1021030-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126150
Availability details for
Quantity
  • Availability
  • Add to Cart