ENA390347369 Enamine

4-chloro-1-hydroxynaphthalene-2-carbaldehyde

  • MDL number MFCD21233452
  • Cas Number : 58132-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126160
Availability details for
Quantity
  • Availability
  • Add to Cart