ENA390347371 Enamine

4-(2-chloroethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD11164691
  • Cas Number : 1094336-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124990
Availability details for
Quantity
  • Availability
  • Add to Cart