ENA390347372 Enamine

2-{4-[2-(3,4-dimethylpyrrolidin-1-yl)ethoxy]phenyl}ethan-1-amine

  • MDL number MFCD22848929
  • Cas Number : 1432681-59-8
  • Molecular Weight : 262.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-(2-(3,4-dimethylpyrrolidin-1-yl)ethoxy)phenyl)ethan-1-amine
Inchi Code 1S/C16H26N2O/c1-13-11-18(12-14(13)2)9-10-19-16-5-3-15(4-6-16)7-8-17/h3-6,13-14H,7-12,17H2,1-2H3
Inchi Key CSSVUAGVDPMDRF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126164
Availability details for
Quantity
  • Availability
  • Add to Cart