ENA390347378 Enamine

4-carbamoyl-2-(3-cyclopentylpropanamido)butanoic acid

  • MDL number MFCD12444867
  • Cas Number : 1105053-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124994
Availability details for
Quantity
  • Availability
  • Add to Cart