ENA390347384 Enamine

[3-(1H-imidazol-2-yl)phenyl]methanamine dihydrochloride

  • MDL number MFCD08668832
  • Cas Number : 1432679-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125012

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125012
Availability details for
Quantity
  • Availability
  • Add to Cart