ENA390347397 Enamine

1-bromo-2-(ethylsulfanyl)ethane

  • MDL number MFCD00236595
  • Cas Number : 35420-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126226

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126226
Availability details for
Quantity
  • Availability
  • Add to Cart