ENA390347406 Enamine

[4-(2-methoxyethyl)phenyl]methanol

  • MDL number MFCD19596968
  • Cas Number : 886531-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126238

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126238
Availability details for
Quantity
  • Availability
  • Add to Cart