ENA390347407 Enamine

3-{[(5-methylpyrimidin-2-yl)amino]methyl}benzoic acid

  • MDL number MFCD19119332
  • Cas Number : 1273847-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126239
Availability details for
Quantity
  • Availability
  • Add to Cart