ENA390347411 Enamine

N-(4-fluorophenyl)-2-nitroaniline

  • MDL number MFCD03414600
  • Cas Number : 322-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126245
Availability details for
Quantity
  • Availability
  • Add to Cart