ENA390347417 Enamine

1-methyl-1H-imidazole-2-carbothioamide

  • MDL number MFCD05662456
  • Cas Number : 885281-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125046

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125046
Availability details for
Quantity
  • Availability
  • Add to Cart