ENA390347420 Enamine

3-nitro-5,6,7,8-tetrahydro-1,6-naphthyridine dihydrochloride

  • MDL number MFCD08752594
  • Cas Number : 1187928-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126256
Availability details for
Quantity
  • Availability
  • Add to Cart