ENA390347422 Enamine

2,2,2-trifluoro-1-[3-(trifluoromethyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD16652261
  • Cas Number : 1228879-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126258
Availability details for
Quantity
  • Availability
  • Add to Cart