ENA390347432 Enamine

4-chloro-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD11219718
  • Cas Number : 66041-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126296
Availability details for
Quantity
  • Availability
  • Add to Cart