ENA390347433 Enamine

6-amino-1-(1-{bicyclo[2.2.1]heptan-2-yl}ethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD11156256
  • Cas Number : 1132676-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248...250
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126297
Availability details for
Quantity
  • Availability
  • Add to Cart