ENA390347437 Enamine

2-(chloromethyl)-1-benzothiophene

  • MDL number MFCD08060638
  • Cas Number : 2076-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126302
Availability details for
Quantity
  • Availability
  • Add to Cart