ENA390347446 Enamine

2,5,5-trimethylmorpholine

  • MDL number MFCD09722891
  • Cas Number : 938374-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125055

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125055
Availability details for
Quantity
  • Availability
  • Add to Cart