ENA390347449 Enamine

(2S)-1-(morpholin-4-yl)propan-2-ol

  • MDL number MFCD23701950
  • Cas Number : 65617-17-6
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-1-morpholinopropan-2-ol
Inchi Code 1S/C7H15NO2/c1-7(9)6-8-2-4-10-5-3-8/h7,9H,2-6H2,1H3/t7-/m0/s1
Inchi Key YAXQOLYGKLGQKA-ZETCQYMHSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126315

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-126315
pictograms GHS05,GHS06
hstatements H301,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart