ENA390347452 Enamine

2-(3,4-dichlorophenyl)propan-1-ol

  • MDL number MFCD21775292
  • Cas Number : 1432678-12-0
  • Molecular Weight : 205.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126319
Availability details for
Quantity
  • Availability
  • Add to Cart