ENA390347458 Enamine

(2S)-1-(1-methyl-2-oxo-1,2-dihydropyridine-4-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD16377151
  • Cas Number : 1309019-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126326
Availability details for
Quantity
  • Availability
  • Add to Cart