ENA390347459 Enamine

2-methyl-6-(trifluoromethyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD09804425
  • Cas Number : 1016703-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126329
Availability details for
Quantity
  • Availability
  • Add to Cart