ENA390347462 Enamine

3-fluoro-2-(methylamino)benzamide

  • MDL number MFCD16836276
  • Cas Number : 1247217-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126353
Availability details for
Quantity
  • Availability
  • Add to Cart