ENA390347471 Enamine

1-[(2-bromophenyl)methyl]piperidin-4-one

  • MDL number MFCD09933043
  • Cas Number : 1016770-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125059
Availability details for
Quantity
  • Availability
  • Add to Cart