ENA390347474 Enamine

1-N,1-N,2-trimethylbenzene-1,3-diamine

  • MDL number MFCD17284897
  • Cas Number : 36714-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126370

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-126370
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart