ENA390347502 Enamine

1-[4-(4-chlorophenyl)piperazin-1-yl]-2-methylpropan-2-amine

  • MDL number MFCD13327248
  • Cas Number : 1306199-31-4
  • Molecular Weight : 267.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126461
Availability details for
Quantity
  • Availability
  • Add to Cart